The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002597.4 | NP_002588.3 | 2 - 3 | 131 | 121 |
XM_011509603.2 | XP_011507905.1 | 1 - 2 | 1149 | 121 | |
GenBank mRNA | EL945964.1 | - | 2 - 3 | 135 | 121 |
M33478.1 | - | 2 - 3 | 108 | 121 |