The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003571.3 | NP_003562.1 | 1 - 2 | 578 | 89 |
XM_017007315.1 | XP_016862804.1 | 1 - 2 | 2691 | 89 | |
GenBank mRNA | BC113518.1 | - | 1 - 2 | 564 | 89 |
BC113520.1 | - | 1 - 2 | 564 | 89 | |
BM696550.1 | - | 1 - 2 | 578 | 89 | |
U48224.1 | - | 1 - 2 | 489 | 89 |