The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003571.3 | NP_003562.1 | 2 - 3 | 663 | 131 |
XM_017007315.1 | XP_016862804.1 | 2 - 3 | 2776 | 131 | |
GenBank mRNA | BC113518.1 | - | 2 - 3 | 649 | 131 |
BC113520.1 | - | 2 - 3 | 649 | 131 | |
U48224.1 | - | 2 - 3 | 574 | 131 |