Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLITRK2
Entrez Gene ID
84631
Gene Name
SLIT and NTRK like family member 2
Gene Aliases
CXorf2, SLITL1
Chromosome Location
Chr.X: 145817829 - 145829856 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005262342.3 XP_005262399.1 5 - 5 8699 68
XM_005262343.3 XP_005262400.1 5 - 5 8709 68
XM_005262344.3 XP_005262401.1 5 - 5 8687 68
XM_005262345.3 XP_005262402.1 4 - 4 8550 68

Gene Symbol
TMEM257
Entrez Gene ID
9142
Gene Name
transmembrane protein 257
Gene Aliases
CXorf1
Chromosome Location
Chr.X: 145827410 - 145829856 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004709.2 NP_004700.1 1 - 1 2123 68
GenBank mRNA AF007148.1 - 1 - 1 1574 68
AL134413.1 - 1 - 1 317 68
Y08902.1 - 1 - 1 2123 68

View Full Product Details