The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318015.1 | NP_001304944.1 | 2 - 3 | 438 | 92 |
NM_001318016.1 | NP_001304945.1 | 2 - 3 | 273 | 92 | |
NM_001318020.1 | NP_001304949.1 | 2 - 3 | 273 | 92 | |
NM_001318023.1 | NP_001304952.1 | 2 - 3 | 273 | 92 | |
NM_032799.4 | NP_116188.2 | 2 - 3 | 273 | 92 | |
XM_011519116.2 | XP_011517418.1 | 2 - 3 | 447 | 92 | |
GenBank mRNA | AK027430.1 | - | 2 - 3 | 266 | 92 |
AK075332.1 | - | 2 - 3 | 266 | 92 | |
AK092843.1 | - | 1 - 2 | 1690 | 92 | |
AK315746.1 | - | 2 - 3 | 269 | 92 | |
BC009280.2 | - | 2 - 3 | 250 | 92 | |
BC048251.1 | - | 2 - 3 | 419 | 92 | |
BG767826.1 | - | 2 - 3 | 247 | 92 | |
BM043458.1 | - | 2 - 3 | 241 | 92 | |
BM745076.1 | - | 2 - 3 | 265 | 92 | |
BU506687.1 | - | 2 - 3 | 274 | 92 |