The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135746.1 | NP_001129218.1 | 1 - 2 | 132 | 92 |
XM_005272382.2 | XP_005272439.1 | 1 - 2 | 103 | 92 | |
GenBank mRNA | AK225837.1 | - | 1 - 2 | 58 | 92 |
BC013952.2 | - | 1 - 2 | 46 | 92 | |
BC013964.2 | - | 1 - 2 | 46 | 92 | |
BX537529.1 | - | 1 - 2 | 48 | 92 | |
DA207321.1 | - | 1 - 2 | 85 | 92 | |
DB452586.1 | - | 1 - 2 | 133 | 92 |