The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020949.2 | NP_066000.2 | 3 - 4 | 856 | 95 |
GenBank mRNA | AB046833.1 | - | 2 - 3 | 307 | 95 |
AK122655.1 | - | 3 - 4 | 851 | 95 | |
BC022968.1 | - | 2 - 3 | 472 | 95 |