The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020949.2 | NP_066000.2 | 4 - 5 | 1082 | 77 |
GenBank mRNA | AB046833.1 | - | 3 - 4 | 533 | 77 |
AK122655.1 | - | 4 - 5 | 1077 | 77 | |
BC022968.1 | - | 3 - 4 | 698 | 77 |