Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NIPAL3
Entrez Gene ID
57185
Gene Name
NIPA like domain containing 3
Gene Aliases
DJ462O23.2, NPAL3
Chromosome Location
Chr.1: 24413360 - 24472983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322854.1 NP_001309783.1 10 - 11 1478 73
NM_001322855.1 NP_001309784.1 11 - 12 1620 73
NM_001322856.1 NP_001309785.1 11 - 12 1577 73
NM_001322857.1 NP_001309786.1 10 - 11 1398 73
NM_001322858.1 NP_001309787.1 11 - 12 1495 73
NM_001322859.1 NP_001309788.1 8 - 9 1104 73
NM_001322860.1 NP_001309789.1 7 - 8 1044 73
NM_001322861.1 NP_001309790.1 7 - 8 754 73
NM_001322862.1 NP_001309791.1 11 - 12 1455 73
NM_001322863.1 NP_001309792.1 10 - 11 1427 73
NM_001322864.1 NP_001309793.1 9 - 10 995 73
NM_001322866.1 NP_001309795.1 10 - 11 1318 73
NM_020448.4 NP_065181.1 10 - 11 1340 73
NR_136506.1 9 - 10 1248 73
NR_136508.1 9 - 10 1209 73
XM_011541808.2 XP_011540110.1 9 - 10 1255 73
XM_017001868.1 XP_016857357.1 9 - 10 990 73
XM_017001869.1 XP_016857358.1 10 - 11 1325 73
XM_017001870.1 XP_016857359.1 11 - 12 1590 73
GenBank mRNA AK312433.1 - 10 - 11 1294 73
BC063583.1 - 10 - 11 1270 73
BX538110.1 - 4 - 5 368 73
BX640883.1 - 11 - 12 1402 73
U90918.1 - 10 - 11 1231 73

View Full Product Details