Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPHN
Entrez Gene ID
10243
Gene Name
gephyrin
Gene Aliases
GEPH, GPH, GPHRYN, HKPX1, MOCODC
Chromosome Location
Chr.14: 66507407 - 67735831 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024218.1 NP_001019389.1 6 - 7 1577 89
NM_020806.4 NP_065857.1 6 - 7 1577 89
XM_005267254.3 XP_005267311.1 6 - 7 872 89
XM_011536340.2 XP_011534642.1 7 - 8 1089 89
XM_011536342.2 XP_011534644.1 6 - 7 1050 89
XM_011536343.2 XP_011534645.1 7 - 8 1089 89
XM_011536344.2 XP_011534646.1 7 - 8 1089 89
XM_011536345.2 XP_011534647.1 7 - 8 1089 89
XM_011536346.2 XP_011534648.1 6 - 7 851 89
XM_011536347.2 XP_011534649.1 7 - 8 575 89
XM_017020913.1 XP_016876402.1 7 - 8 1089 89
XM_017020914.1 XP_016876403.1 7 - 8 1089 89
XM_017020915.1 XP_016876404.1 7 - 8 1089 89
XM_017020916.1 XP_016876405.1 6 - 7 1050 89
XM_017020917.1 XP_016876406.1 7 - 8 1089 89
XM_017020918.1 XP_016876407.1 7 - 8 1089 89
XM_017020919.1 XP_016876408.1 6 - 7 1050 89
XM_017020920.1 XP_016876409.1 7 - 8 1012 89
XM_017020921.1 XP_016876410.1 7 - 8 1152 89
XM_017020922.1 XP_016876411.1 8 - 9 618 89
XM_017020923.1 XP_016876412.1 7 - 8 1152 89
XM_017020924.1 XP_016876413.1 7 - 8 1089 89
XM_017020925.1 XP_016876414.1 7 - 8 1089 89
XM_017020926.1 XP_016876415.1 7 - 8 1089 89
GenBank mRNA AB037806.1 - 6 - 7 1577 89
AF272663.1 - 6 - 7 1577 89
AJ272033.1 - 6 - 7 456 89
AJ272343.1 - 6 - 7 496 89
AK294532.1 - 7 - 8 519 89
AK295577.1 - 4 - 5 354 89
AK297447.1 - 5 - 6 378 89
AK315569.1 - 6 - 7 826 89

View Full Product Details