The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024057.3 | NP_076962.2 | 2 - 3 | 307 | 87 |
GenBank mRNA | AF514994.1 | - | 1 - 2 | 161 | 87 |
AK026271.1 | - | 1 - 2 | 142 | 87 | |
AK296733.1 | - | 2 - 3 | 265 | 87 | |
AK310619.1 | - | 2 - 3 | 267 | 87 | |
BC000861.2 | - | 1 - 2 | 225 | 87 | |
DC420994.1 | - | 2 - 3 | 308 | 87 |