Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUP37
Entrez Gene ID
79023
Gene Name
nucleoporin 37
Gene Aliases
p37
Chromosome Location
Chr.12: 102074189 - 102120139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024057.3 NP_076962.2 4 - 5 502 161
GenBank mRNA AF514994.1 - 3 - 4 356 161
AK026271.1 - 3 - 4 337 161
AK296733.1 - 4 - 5 460 161
AK310619.1 - 4 - 5 462 161
BC000861.2 - 3 - 4 420 161
DC420994.1 - 4 - 5 503 161

View Full Product Details