Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDH1
Entrez Gene ID
622
Gene Name
3-hydroxybutyrate dehydrogenase, type 1
Gene Aliases
BDH, SDR9C1
Chromosome Location
Chr.3: 197509783 - 197573323 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004051.4 NP_004042.1 4 - 5 524 61
NM_203314.2 NP_976059.1 5 - 6 676 61
NM_203315.2 NP_976060.1 4 - 5 460 61
XM_005269352.3 XP_005269409.1 4 - 5 340 61
XM_011513067.2 XP_011511369.1 3 - 4 1889 61
XM_017007007.1 XP_016862496.1 4 - 5 1968 61
XM_017007008.1 XP_016862497.1 5 - 6 2588 61
XM_017007009.1 XP_016862498.1 6 - 7 1543 61
XM_017007010.1 XP_016862499.1 5 - 6 1391 61
XM_017007011.1 XP_016862500.1 7 - 8 1601 61
XM_017007012.1 XP_016862501.1 6 - 7 1449 61
GenBank mRNA BC005844.2 - 4 - 5 488 61
BC011964.1 - 4 - 5 473 61
BC019317.1 - 4 - 5 488 61
BE782251.1 - - 584 61
BI601294.1 - 5 - 6 564 61
BQ897235.1 - 4 - 5 521 61
M93107.1 - 3 - 4 274 61

View Full Product Details