Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GZF1
Entrez Gene ID
64412
Gene Name
GDNF inducible zinc finger protein 1
Gene Aliases
ZBTB23, ZNF336
Chromosome Location
Chr.20: 23361585 - 23373062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317012.1 NP_001303941.1 6 - 7 2013 90
NM_022482.4 NP_071927.1 5 - 6 1916 90
XM_011529321.2 XP_011527623.1 5 - 6 1888 90
XM_011529322.1 XP_011527624.1 5 - 6 1849 90
GenBank mRNA AB100265.1 - 4 - 5 1789 90
AK025447.1 - 4 - 5 833 90
AK056159.1 - 4 - 5 1410 90
AK056477.1 - 5 - 6 1866 90
AK289814.1 - 6 - 7 1928 90
AK293942.1 - 3 - 4 386 90
AK295362.1 - 4 - 5 513 90
AK314599.1 - 5 - 6 1866 90

View Full Product Details