The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042467.2 | NP_001035932.1 | 6 - 7 | 915 | 98 |
NM_024101.6 | NP_077006.1 | 6 - 7 | 915 | 98 | |
NR_104019.1 | 6 - 7 | 915 | 98 | ||
XM_006712739.1 | XP_006712802.1 | 6 - 7 | 883 | 98 | |
XM_011511812.1 | XP_011510114.1 | 3 - 4 | 2575 | 98 | |
XM_017004893.1 | XP_016860382.1 | 6 - 7 | 883 | 98 | |
XM_017004894.1 | XP_016860383.1 | 6 - 7 | 883 | 98 | |
GenBank mRNA | AK022207.1 | - | 6 - 7 | 98 | |
AK056867.1 | - | 4 - 5 | 1049 | 98 | |
AK292929.1 | - | 6 - 7 | 883 | 98 | |
AK296745.1 | - | 6 - 7 | 884 | 98 | |
AK310821.1 | - | 6 - 7 | 883 | 98 | |
AY358857.1 | - | 2 - 3 | 737 | 98 | |
BC001653.2 | - | 6 - 7 | 847 | 98 | |
BC051269.1 | - | 6 - 7 | 857 | 98 |