The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002755.3 | NP_002746.1 | 2 - 3 | 762 | 67 |
XM_011521783.2 | XP_011520085.1 | 2 - 3 | 419 | 67 | |
XM_017022411.1 | XP_016877900.1 | 2 - 3 | 713 | 67 | |
XM_017022412.1 | XP_016877901.1 | 2 - 3 | 299 | 67 | |
GenBank mRNA | AK291500.1 | - | 2 - 3 | 723 | 67 |
AK294322.1 | - | 2 - 3 | 273 | 67 | |
AK312356.1 | - | 2 - 3 | 669 | 67 | |
BC137459.1 | - | 2 - 3 | 350 | 67 | |
BC139729.1 | - | 2 - 3 | 346 | 67 | |
L05624.1 | - | 2 - 3 | 320 | 67 | |
L11284.1 | - | 2 - 3 | 359 | 67 |