The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002755.3 | NP_002746.1 | 7 - 8 | 1371 | 106 |
XM_011521783.2 | XP_011520085.1 | 7 - 8 | 1028 | 106 | |
XM_017022411.1 | XP_016877900.1 | 6 - 7 | 1244 | 106 | |
XM_017022412.1 | XP_016877901.1 | 6 - 7 | 830 | 106 | |
XM_017022413.1 | XP_016877902.1 | 3 - 4 | 1439 | 106 | |
GenBank mRNA | AK291500.1 | - | 7 - 8 | 1332 | 106 |
AK294322.1 | - | 7 - 8 | 882 | 106 | |
AK312356.1 | - | 7 - 8 | 1278 | 106 | |
BC137459.1 | - | 7 - 8 | 959 | 106 | |
BC139729.1 | - | 7 - 8 | 955 | 106 | |
L05624.1 | - | 7 - 8 | 929 | 106 | |
L11284.1 | - | 7 - 8 | 968 | 106 |