The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002755.3 | NP_002746.1 | 9 - 10 | 1490 | 86 |
XM_011521783.2 | XP_011520085.1 | 9 - 10 | 1147 | 86 | |
XM_017022411.1 | XP_016877900.1 | 8 - 9 | 1363 | 86 | |
XM_017022412.1 | XP_016877901.1 | 8 - 9 | 949 | 86 | |
XM_017022413.1 | XP_016877902.1 | 5 - 6 | 1558 | 86 | |
GenBank mRNA | AK291500.1 | - | 9 - 10 | 1451 | 86 |
AK294322.1 | - | 9 - 10 | 1001 | 86 | |
AK312356.1 | - | 9 - 10 | 1397 | 86 | |
BC137459.1 | - | 9 - 10 | 1078 | 86 | |
BC139729.1 | - | 9 - 10 | 1074 | 86 | |
L05624.1 | - | 9 - 10 | 1048 | 86 | |
L11284.1 | - | 9 - 10 | 1087 | 86 |