Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NPVF
Entrez Gene ID
64111
Gene Name
neuropeptide VF precursor
Gene Aliases
C7orf9, RFRP
Chromosome Location
Chr.7: 25224570 - 25228486 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022150.3 NP_071433.3 2 - 3 590 106
GenBank mRNA AF440392.1 - 2 - 3 590 106
BM714303.1 - 2 - 3 535 106

View Full Product Details