Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AKAP14
Entrez Gene ID
158798
Gene Name
A-kinase anchoring protein 14
Gene Aliases
AKAP28, PRKA14
Chromosome Location
Chr.X: 119895973 - 119920716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC066357.1 - 1 - 2 153 74

View Full Product Details