The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008534.1 | NP_001008534.1 | 2 - 3 | 306 | 91 |
NM_001008535.1 | NP_001008535.1 | 2 - 3 | 306 | 91 | |
NM_178813.5 | NP_848928.1 | 2 - 3 | 306 | 91 | |
GenBank mRNA | AF514780.1 | - | 2 - 3 | 253 | 91 |
AF514781.1 | - | 2 - 3 | 253 | 91 | |
AF514782.1 | - | 2 - 3 | 253 | 91 | |
BC066357.1 | - | 3 - 4 | 443 | 91 | |
BG208671.1 | - | 1 - 2 | 230 | 91 |