Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM243
Entrez Gene ID
79161
Gene Name
transmembrane protein 243
Gene Aliases
C7orf23, MM-TRAG, MMTRAG
Chromosome Location
Chr.7: 87195593 - 87220603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005250586.3 XP_005250643.1 3 - 4 560 97
XM_005250587.4 XP_005250644.1 3 - 4 694 97
XM_005250588.3 XP_005250645.1 3 - 4 392 97
XM_005250589.3 XP_005250646.1 3 - 4 560 97
XM_017012616.1 XP_016868105.1 2 - 3 981 97
XM_017012617.1 XP_016868106.1 3 - 4 683 97
XM_017012618.1 XP_016868107.1 3 - 4 427 97
GenBank mRNA AK313794.1 - 2 - 3 362 97
BC002837.2 - 2 - 3 344 97
BT006973.1 - 2 - 3 132 97
CB993361.1 - 3 - 4 601 97

View Full Product Details