The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015957.3 | NP_057041.2 | 2 - 3 | 290 | 72 |
XM_011520154.2 | XP_011518456.1 | 3 - 4 | 363 | 72 | |
XM_017017875.1 | XP_016873364.1 | 3 - 4 | 337 | 72 | |
GenBank mRNA | AF132963.1 | - | 2 - 3 | 240 | 72 |
AK225249.1 | - | 2 - 3 | 222 | 72 | |
AK292648.1 | - | 2 - 3 | 258 | 72 | |
AK302225.1 | - | 3 - 4 | 310 | 72 | |
BC008440.1 | - | 2 - 3 | 257 | 72 | |
BC009077.1 | - | 2 - 3 | 204 | 72 | |
BC017594.2 | - | 2 - 3 | 199 | 72 | |
BP200131.1 | - | 2 - 3 | 379 | 72 | |
DA729016.1 | - | 2 - 3 | 271 | 72 |