Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSGALNACT1
Entrez Gene ID
55790
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
CSGalNAcT-1, ChGn, beta4GalNAcT
Chromosome Location
Chr.8: 19404161 - 19757940 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130518.1 NP_001123990.1 5 - 6 1912 91
NM_018371.4 NP_060841.5 5 - 6 1502 91
XM_006716358.2 XP_006716421.1 4 - 5 1793 91
XM_006716359.2 XP_006716422.1 5 - 6 2162 91
XM_006716360.2 XP_006716423.1 6 - 7 2310 91
XM_006716361.2 XP_006716424.1 5 - 6 2057 91
XM_006716362.2 XP_006716425.1 4 - 5 1380 91
XM_006716363.1 XP_006716426.1 4 - 5 1552 91
XM_006716364.3 XP_006716427.1 3 - 4 1236 91
XM_011544578.1 XP_011542880.1 7 - 8 2429 91
XM_011544579.1 XP_011542881.1 6 - 7 2281 91
XM_011544580.1 XP_011542882.1 5 - 6 2159 91
XM_011544582.1 XP_011542884.1 5 - 6 2167 91
XM_011544583.1 XP_011542885.1 3 - 4 1346 91
XM_011544584.1 XP_011542886.1 3 - 4 1230 91
XM_011544585.2 XP_011542887.1 2 - 3 313 91
XM_017013623.1 XP_016869112.1 8 - 9 2479 91
XM_017013624.1 XP_016869113.1 5 - 6 2118 91
XM_017013625.1 XP_016869114.1 3 - 4 1395 91
XM_017013626.1 XP_016869115.1 4 - 5 1325 91
XM_017013627.1 XP_016869116.1 5 - 6 3719 91
XM_017013628.1 XP_016869117.1 4 - 5 3579 91
XM_017013629.1 XP_016869118.1 5 - 6 1510 91
XM_017013630.1 XP_016869119.1 5 - 6 1423 91
XM_017013631.1 XP_016869120.1 4 - 5 1384 91
XM_017013632.1 XP_016869121.1 5 - 6 1493 91
XM_017013633.1 XP_016869122.1 5 - 6 1480 91
XM_017013634.1 XP_016869123.1 5 - 6 1477 91
XM_017013635.1 XP_016869124.1 5 - 6 1485 91
XM_017013636.1 XP_016869125.1 6 - 7 1628 91
XM_017013637.1 XP_016869126.1 5 - 6 1485 91
XM_017013638.1 XP_016869127.1 6 - 7 1670 91
XM_017013639.1 XP_016869128.1 4 - 5 1318 91
GenBank mRNA AB071403.1 - 2 - 3 872 91
AB081516.1 - 3 - 4 1190 91
AK314625.1 - 4 - 5 1261 91
AY358441.1 - 5 - 6 1502 91
BX649103.1 - 5 - 6 1862 91

View Full Product Details