Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPN3
Entrez Gene ID
5774
Gene Name
protein tyrosine phosphatase, non-receptor type 3
Gene Aliases
PTP-H1, PTPH1
Chromosome Location
Chr.9: 109375694 - 109538395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145368.1 NP_001138840.1 2 - 3 252 69
NM_002829.3 NP_002820.3 2 - 3 252 69
XM_006717197.3 XP_006717260.1 2 - 3 361 69
XM_006717199.3 XP_006717262.1 2 - 3 386 69
XM_006717201.3 XP_006717264.1 2 - 3 272 69
XM_006717204.3 XP_006717267.1 2 - 3 252 69
XM_011518888.2 XP_011517190.1 2 - 3 322 69
XM_011518889.2 XP_011517191.1 2 - 3 992 69
XM_017014955.1 XP_016870444.1 2 - 3 344 69
XM_017014956.1 XP_016870445.1 2 - 3 279 69
XM_017014957.1 XP_016870446.1 2 - 3 252 69
GenBank mRNA AK295875.1 - 2 - 3 251 69
AK304434.1 - 2 - 3 252 69
AK316066.1 - 2 - 3 252 69
BC126117.1 - 1 - 2 162 69
BC143848.1 - 1 - 2 162 69
DQ104439.1 - 1 - 2 140 69
M64572.1 - 1 - 2 163 69

View Full Product Details