Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPN3
Entrez Gene ID
5774
Gene Name
protein tyrosine phosphatase, non-receptor type 3
Gene Aliases
PTP-H1, PTPH1
Chromosome Location
Chr.9: 109375694 - 109538395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145368.1 NP_001138840.1 5 - 6 481 111
NM_002829.3 NP_002820.3 5 - 6 481 111
XM_006717197.3 XP_006717260.1 5 - 6 590 111
XM_006717199.3 XP_006717262.1 5 - 6 615 111
XM_011518888.2 XP_011517190.1 5 - 6 551 111
XM_011518889.2 XP_011517191.1 5 - 6 1221 111
XM_017014955.1 XP_016870444.1 5 - 6 573 111
GenBank mRNA AK304434.1 - 5 - 6 481 111
AK316066.1 - 5 - 6 481 111
BC126117.1 - 4 - 5 391 111
BC143848.1 - 4 - 5 391 111
DQ104439.1 - 4 - 5 369 111
M64572.1 - 4 - 5 392 111

View Full Product Details