The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173598.4 | NP_775869.3 | 14 - 15 | 2184 | 77 |
XM_011538224.2 | XP_011536526.1 | 14 - 15 | 2513 | 77 | |
XM_011538225.2 | XP_011536527.1 | 14 - 15 | 2978 | 77 | |
XM_011538228.2 | XP_011536530.1 | 12 - 13 | 1343 | 77 | |
XM_017019208.1 | XP_016874697.1 | 14 - 15 | 2519 | 77 | |
XM_017019210.1 | XP_016874699.1 | 9 - 10 | 969 | 77 | |
GenBank mRNA | AY345972.1 | - | 12 - 13 | 1853 | 77 |
BC107106.1 | - | 12 - 13 | 1848 | 77 | |
BC107107.1 | - | 12 - 13 | 1848 | 77 | |
BC127603.1 | - | 12 - 13 | 1853 | 77 |