Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHF1
Entrez Gene ID
5252
Gene Name
PHD finger protein 1
Gene Aliases
MTF2L2, PCL1, PHF2, TDRD19C, hPHF1
Chromosome Location
Chr.6: 33410751 - 33416453 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002636.4 NP_002627.1 13 - 14 1542 117
NM_024165.2 NP_077084.1 14 - 15 1648 117
NR_027692.1 14 - 15 1637 117
XM_011514662.1 XP_011512964.1 14 - 15 1565 117
XM_011514663.1 XP_011512965.1 14 - 15 1562 117
XM_011514664.1 XP_011512966.1 13 - 14 1469 117
XM_011514665.1 XP_011512967.1 14 - 15 1585 117
XM_011514666.1 XP_011512968.1 14 - 15 1582 117
XM_011514669.1 XP_011512971.1 13 - 14 1375 117
XM_017010939.1 XP_016866428.1 12 - 13 1253 117
GenBank mRNA AF029678.1 - 13 - 14 1362 117
AF052205.1 - 14 - 15 1464 117
AK225913.1 - 13 - 14 1650 117
AK316499.1 - 14 - 15 1588 117
BC008834.2 - 14 - 15 1616 117

View Full Product Details