The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013432.4 | NP_038460.4 | 4 - 5 | 464 | 74 |
XM_011517050.2 | XP_011515352.1 | 4 - 5 | 523 | 74 | |
GenBank mRNA | BC008782.2 | - | 3 - 4 | 436 | 74 |
CN366457.1 | - | 4 - 5 | 488 | 74 |