Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INPP5D
Entrez Gene ID
3635
Gene Name
inositol polyphosphate-5-phosphatase D
Gene Aliases
SHIP, SHIP-1, SHIP1, SIP-145, hp51CN, p150Ship
Chromosome Location
Chr.2: 233059967 - 233207903 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017915.2 NP_001017915.1 19 - 20 2675 63
NM_005541.4 NP_005532.2 19 - 20 2672 63
XM_017004004.1 XP_016859493.1 19 - 20 2251 63
GenBank mRNA BC099920.1 - 19 - 20 2283 63
BC113580.1 - 19 - 20 2220 63
BC113582.1 - 19 - 20 2220 63
U50040.1 - 14 - 15 1540 63
U50041.1 - 19 - 20 2284 63
U53470.1 - 19 - 20 2293 63
U57650.1 - 19 - 20 2672 63
U84400.1 - 19 - 20 2244 63
X98429.1 - 19 - 20 2300 63

View Full Product Details