The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017915.2 | NP_001017915.1 | 1 - 2 | 641 | 68 |
NM_005541.4 | NP_005532.2 | 1 - 2 | 641 | 68 | |
XM_017004004.1 | XP_016859493.1 | 1 - 2 | 264 | 68 | |
GenBank mRNA | AK298651.1 | - | 1 - 2 | 270 | 68 |
AK309644.1 | - | 1 - 2 | 266 | 68 | |
BC099920.1 | - | 1 - 2 | 252 | 68 | |
BC113580.1 | - | 1 - 2 | 189 | 68 | |
BC113582.1 | - | 1 - 2 | 189 | 68 | |
BE247227.1 | - | 1 - 2 | 286 | 68 | |
BM800110.1 | - | - | 278 | 68 | |
U50041.1 | - | 1 - 2 | 250 | 68 | |
U53470.1 | - | 1 - 2 | 259 | 68 | |
U57650.1 | - | 1 - 2 | 641 | 68 | |
U84400.1 | - | 1 - 2 | 210 | 68 | |
X98429.1 | - | 1 - 2 | 269 | 68 |