Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INPP5D
Entrez Gene ID
3635
Gene Name
inositol polyphosphate-5-phosphatase D
Gene Aliases
SHIP, SHIP-1, SHIP1, SIP-145, hp51CN, p150Ship
Chromosome Location
Chr.2: 233059967 - 233207903 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017915.2 NP_001017915.1 21 - 22 2877 64
NM_005541.4 NP_005532.2 21 - 22 2874 64
XM_017004004.1 XP_016859493.1 21 - 22 2453 64
GenBank mRNA BC099920.1 - 21 - 22 2485 64
BC113580.1 - 21 - 22 2422 64
BC113582.1 - 21 - 22 2422 64
U50040.1 - 16 - 17 1742 64
U50041.1 - 21 - 22 2486 64
U53470.1 - 21 - 22 2495 64
U57650.1 - 21 - 22 2874 64
U84400.1 - 21 - 22 2446 64
X98429.1 - 21 - 22 2502 64

View Full Product Details