The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017915.2 | NP_001017915.1 | 2 - 3 | 712 | 98 |
NM_005541.4 | NP_005532.2 | 2 - 3 | 712 | 98 | |
XM_017004004.1 | XP_016859493.1 | 2 - 3 | 335 | 98 | |
GenBank mRNA | AK298651.1 | - | 2 - 3 | 341 | 98 |
AK309644.1 | - | 2 - 3 | 337 | 98 | |
BC099920.1 | - | 2 - 3 | 323 | 98 | |
BC113580.1 | - | 2 - 3 | 260 | 98 | |
BC113582.1 | - | 2 - 3 | 260 | 98 | |
U50041.1 | - | 2 - 3 | 321 | 98 | |
U53470.1 | - | 2 - 3 | 330 | 98 | |
U57650.1 | - | 2 - 3 | 712 | 98 | |
U84400.1 | - | 2 - 3 | 281 | 98 | |
X98429.1 | - | 2 - 3 | 340 | 98 |