The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000722.3 | NP_000713.2 | 10 - 11 | 1232 | 84 |
XM_005250570.2 | XP_005250627.1 | 10 - 11 | 1339 | 84 | |
XM_005250572.2 | XP_005250629.1 | 10 - 11 | 1339 | 84 | |
XM_005250573.2 | XP_005250630.1 | 10 - 11 | 1336 | 84 | |
XM_005250574.2 | XP_005250631.1 | 10 - 11 | 1337 | 84 | |
XM_006716118.2 | XP_006716181.1 | 10 - 11 | 1341 | 84 | |
XM_006716119.3 | XP_006716182.1 | 10 - 11 | 3124 | 84 | |
XM_006716120.3 | XP_006716183.1 | 10 - 11 | 2566 | 84 | |
XM_011516570.2 | XP_011514872.1 | 10 - 11 | 1347 | 84 | |
XM_011516571.2 | XP_011514873.1 | 10 - 11 | 1342 | 84 | |
XM_011516572.2 | XP_011514874.1 | 10 - 11 | 1339 | 84 | |
XM_017012588.1 | XP_016868077.1 | 10 - 11 | 2565 | 84 | |
GenBank mRNA | BC117468.1 | - | 10 - 11 | 916 | 84 |
BC117470.1 | - | 10 - 11 | 916 | 84 | |
M76559.1 | - | 10 - 11 | 919 | 84 |