The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000722.3 | NP_000713.2 | 1 - 2 | 441 | 105 |
NM_001302890.1 | NP_001289819.1 | 1 - 2 | 441 | 105 | |
XM_005250570.2 | XP_005250627.1 | 1 - 2 | 548 | 105 | |
XM_005250572.2 | XP_005250629.1 | 1 - 2 | 548 | 105 | |
XM_005250573.2 | XP_005250630.1 | 1 - 2 | 545 | 105 | |
XM_005250574.2 | XP_005250631.1 | 1 - 2 | 546 | 105 | |
XM_006716118.2 | XP_006716181.1 | 1 - 2 | 550 | 105 | |
XM_011516570.2 | XP_011514872.1 | 1 - 2 | 556 | 105 | |
XM_011516571.2 | XP_011514873.1 | 1 - 2 | 551 | 105 | |
XM_011516572.2 | XP_011514874.1 | 1 - 2 | 548 | 105 | |
GenBank mRNA | BC051339.1 | - | 1 - 2 | 351 | 105 |
BC117468.1 | - | 1 - 2 | 125 | 105 | |
BC117470.1 | - | 1 - 2 | 125 | 105 | |
BP258070.1 | - | 1 - 2 | 440 | 105 | |
BU501177.1 | - | 1 - 2 | 350 | 105 | |
M76559.1 | - | 1 - 2 | 128 | 105 |