Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA2D1
Entrez Gene ID
781
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 1
Gene Aliases
CACNA2, CACNL2A, CCHL2A, LINC01112, lncRNA-N3
Chromosome Location
Chr.7: 81946444 - 82443921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000722.3 NP_000713.2 31 - 32 2847 69
XM_005250570.2 XP_005250627.1 31 - 32 2990 69
XM_005250572.2 XP_005250629.1 31 - 32 2939 69
XM_005250573.2 XP_005250630.1 30 - 31 2930 69
XM_005250574.2 XP_005250631.1 30 - 31 2916 69
XM_006716118.2 XP_006716181.1 32 - 33 3013 69
XM_006716119.3 XP_006716182.1 32 - 33 4796 69
XM_006716120.3 XP_006716183.1 32 - 33 4238 69
XM_006716121.3 XP_006716184.1 14 - 15 1862 69
XM_011516570.2 XP_011514872.1 32 - 33 3019 69
XM_011516571.2 XP_011514873.1 32 - 33 2999 69
XM_011516572.2 XP_011514874.1 31 - 32 2975 69
XM_017012588.1 XP_016868077.1 31 - 32 4180 69
GenBank mRNA AK299838.1 - 3 - 4 520 69
BC117468.1 - 31 - 32 2531 69
BC117470.1 - 31 - 32 2531 69
M76559.1 - 31 - 32 2534 69

View Full Product Details