Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAD1
Entrez Gene ID
5810
Gene Name
RAD1 checkpoint DNA exonuclease
Gene Aliases
HRAD1, REC1
Chromosome Location
Chr.5: 34905260 - 34915675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002853.3 NP_002844.1 4 - 5 891 94
NR_026591.1 3 - 4 733 94
GenBank mRNA AF011905.1 - 4 - 5 776 94
AF030933.1 - 6 - 7 999 94
AF058392.1 - 3 - 4 562 94
AF058393.1 - 2 - 3 453 94
AF073524.1 - 4 - 5 757 94
AF074717.1 - 4 - 5 701 94
AF076841.1 - 4 - 5 757 94
AF084512.1 - 4 - 5 766 94
AF084513.1 - 3 - 4 657 94
AF090170.1 - 3 - 4 648 94
AJ004974.1 - 4 - 5 725 94
AJ004975.1 - 3 - 4 616 94
AK002112.1 - 4 - 5 894 94
BC006837.1 - 4 - 5 743 94
BC009804.1 - 4 - 5 718 94
BC037857.1 - 4 - 5 1990 94
BT006908.1 - 3 - 4 562 94

View Full Product Details