The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004801.4 | NP_004792.1 | 19 - 20 | 5199 | 66 |
NM_138735.2 | NP_620072.1 | 3 - 4 | 1424 | 66 | |
XM_006712137.3 | XP_006712200.1 | 19 - 20 | 5220 | 66 | |
XM_011533177.2 | XP_011531479.1 | 19 - 20 | 5220 | 66 | |
XM_017005321.1 | XP_016860810.1 | 20 - 21 | 5247 | 66 | |
XM_017005322.1 | XP_016860811.1 | 20 - 21 | 5247 | 66 | |
XM_017005324.1 | XP_016860813.1 | 18 - 19 | 5193 | 66 | |
XM_017005325.1 | XP_016860814.1 | 18 - 19 | 5193 | 66 | |
XM_017005326.1 | XP_016860815.1 | 18 - 19 | 5181 | 66 | |
XM_017005327.1 | XP_016860816.1 | 18 - 19 | 5175 | 66 | |
XM_017005328.1 | XP_016860817.1 | 17 - 18 | 5148 | 66 | |
XM_017005330.1 | XP_016860819.1 | 15 - 16 | 5133 | 66 | |
XM_017005331.1 | XP_016860820.1 | 15 - 16 | 5118 | 66 | |
XM_017005332.1 | XP_016860821.1 | 14 - 15 | 5088 | 66 | |
XM_017005333.1 | XP_016860822.1 | 14 - 15 | 5088 | 66 | |
XM_017005337.1 | XP_016860826.1 | 3 - 4 | 6414 | 66 | |
GenBank mRNA | AB035356.1 | - | 18 - 19 | 3914 | 66 |
AF064842.1 | - | 3 - 4 | 1423 | 66 | |
BC125179.1 | - | 4 - 5 | 480 | 66 | |
BC125180.1 | - | 4 - 5 | 843 | 66 |