Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BBS2
Entrez Gene ID
583
Gene Name
Bardet-Biedl syndrome 2
Gene Aliases
BBS, RP74
Chromosome Location
Chr.16: 56470403 - 56520096 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031885.3 NP_114091.3 9 - 10 1313 69
XM_005256080.2 XP_005256137.1 9 - 10 1312 69
GenBank mRNA AF342736.1 - 9 - 10 1500 69
AK027635.1 - 9 - 10 1241 69
AK289604.1 - 9 - 10 1203 69
BC014140.2 - 9 - 10 1203 69

View Full Product Details