The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145451.2 | NP_001138923.2 | 12 - 13 | 1460 | 91 |
GenBank mRNA | DV080660.1 | - | 2 - 3 | 91 |