The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002203.3 | NP_002194.2 | 3 - 4 | 445 | 86 |
NR_073103.1 | 3 - 4 | 445 | 86 | ||
NR_073104.1 | 3 - 4 | 445 | 86 | ||
NR_073105.1 | 3 - 4 | 445 | 86 | ||
NR_073106.1 | 3 - 4 | 445 | 86 | ||
NR_073107.1 | 2 - 3 | 324 | 86 | ||
GenBank mRNA | AK307952.1 | - | 3 - 4 | 434 | 86 |
BC143505.1 | - | 3 - 4 | 331 | 86 | |
BC143508.1 | - | 3 - 4 | 331 | 86 | |
BC143509.1 | - | 3 - 4 | 331 | 86 | |
BC143511.1 | - | 3 - 4 | 331 | 86 | |
BC143512.1 | - | 2 - 3 | 210 | 86 | |
BP280760.1 | - | 3 - 4 | 447 | 86 | |
X17033.1 | - | 3 - 4 | 350 | 86 |