The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018968.3 | NP_061841.2 | 13 - 14 | 1206 | 79 |
XM_011510366.2 | XP_011508668.1 | 11 - 12 | 1243 | 79 | |
XM_017004361.1 | XP_016859850.1 | 13 - 14 | 1168 | 79 | |
XM_017004362.1 | XP_016859851.1 | 13 - 14 | 1174 | 79 | |
XM_017004363.1 | XP_016859852.1 | 13 - 14 | 1355 | 79 | |
XM_017004364.1 | XP_016859853.1 | 10 - 11 | 4496 | 79 | |
XM_017004365.1 | XP_016859854.1 | 13 - 14 | 1169 | 79 | |
XM_017004366.1 | XP_016859855.1 | 13 - 14 | 1169 | 79 | |
XM_017004367.1 | XP_016859856.1 | 14 - 15 | 1244 | 79 | |
XM_017004373.1 | XP_016859862.1 | 8 - 9 | 1810 | 79 | |
GenBank mRNA | AJ003029.1 | - | 13 - 14 | 1208 | 79 |
BC125251.1 | - | 7 - 8 | 754 | 79 |