The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017742.5 | NP_060212.4 | 10 - 11 | 1752 | 147 |
NR_126534.1 | 10 - 11 | 1752 | 147 | ||
XM_006722493.3 | XP_006722556.1 | 10 - 11 | 1535 | 147 | |
XM_011526044.1 | XP_011524346.1 | 10 - 11 | 2038 | 147 | |
XM_017025802.1 | XP_016881291.1 | 9 - 10 | 1445 | 147 | |
GenBank mRNA | AB051531.1 | - | 9 - 10 | 785 | 147 |
AK098132.1 | - | 3 - 4 | 494 | 147 | |
AL832323.1 | - | 10 - 11 | 916 | 147 | |
BC137435.1 | - | 9 - 10 | 813 | 147 |