The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015285.2 | NP_056100.2 | 15 - 16 | 2971 | 153 |
NM_052834.2 | NP_443066.2 | 15 - 16 | 2971 | 153 | |
XM_006722431.2 | XP_006722494.1 | 15 - 16 | 2903 | 153 | |
XM_011525888.1 | XP_011524190.1 | 15 - 16 | 3013 | 153 | |
XM_017025682.1 | XP_016881171.1 | 15 - 16 | 2876 | 153 | |
XM_017025683.1 | XP_016881172.1 | 5 - 6 | 1364 | 153 | |
GenBank mRNA | AB011113.2 | - | 15 - 16 | 2934 | 153 |
AF188125.1 | - | 14 - 15 | 2760 | 153 | |
AK057141.1 | - | 15 - 16 | 2852 | 153 | |
AY099325.1 | - | 14 - 15 | 2760 | 153 | |
BC131558.1 | - | 3 - 4 | 735 | 153 | |
BC150282.1 | - | 15 - 16 | 2934 | 153 |