Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR7
Entrez Gene ID
23335
Gene Name
WD repeat domain 7
Gene Aliases
TRAG
Chromosome Location
Chr.18: 56651343 - 57029805 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015285.2 NP_056100.2 2 - 3 365 113
NM_052834.2 NP_443066.2 2 - 3 365 113
XM_006722431.2 XP_006722494.1 2 - 3 297 113
XM_011525888.1 XP_011524190.1 2 - 3 407 113
XM_017025682.1 XP_016881171.1 2 - 3 270 113
GenBank mRNA AB011113.2 - 2 - 3 328 113
AK057141.1 - 2 - 3 389 113
AY099325.1 - 1 - 2 154 113
BC150282.1 - 2 - 3 328 113

View Full Product Details