Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR7
Entrez Gene ID
23335
Gene Name
WD repeat domain 7
Gene Aliases
TRAG
Chromosome Location
Chr.18: 56651343 - 57029805 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015285.2 NP_056100.2 22 - 23 3920 90
NM_052834.2 NP_443066.2 21 - 22 3821 90
XM_006722431.2 XP_006722494.1 22 - 23 3852 90
XM_011525888.1 XP_011524190.1 22 - 23 3962 90
XM_017025682.1 XP_016881171.1 21 - 22 3726 90
XM_017025683.1 XP_016881172.1 12 - 13 2313 90
GenBank mRNA AB011113.2 - 22 - 23 3883 90
AF188125.1 - 20 - 21 3610 90
AY099325.1 - 21 - 22 3709 90
BC050352.1 - 8 - 9 1081 90
BC131558.1 - 10 - 11 1684 90
BC150282.1 - 22 - 23 3883 90

View Full Product Details