The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014675.4 | NP_055490.4 | 23 - 24 | 3532 | 89 |
XM_006711058.3 | XP_006711121.1 | 22 - 23 | 3323 | 89 | |
XM_011542468.1 | XP_011540770.1 | 23 - 24 | 3509 | 89 | |
XM_011542470.2 | XP_011540772.1 | 23 - 24 | 3509 | 89 | |
XM_017002913.1 | XP_016858402.1 | 23 - 24 | 3509 | 89 | |
XM_017002914.1 | XP_016858403.1 | 23 - 24 | 3509 | 89 | |
XM_017002915.1 | XP_016858404.1 | 20 - 21 | 3017 | 89 | |
GenBank mRNA | AB007914.2 | - | 21 - 22 | 3171 | 89 |
BC126911.1 | - | 20 - 21 | 3053 | 89 | |
BC126912.1 | - | 20 - 21 | 3053 | 89 | |
DQ139275.1 | - | 10 - 11 | 2889 | 89 |