Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CROCC
Entrez Gene ID
9696
Gene Name
ciliary rootlet coiled-coil, rootletin
Gene Aliases
ROLT
Chromosome Location
Chr.1: 16914082 - 16972979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014675.4 NP_055490.4 18 - 19 2776 83
XM_006711058.3 XP_006711121.1 17 - 18 2567 83
XM_011542468.1 XP_011540770.1 18 - 19 2753 83
XM_011542470.2 XP_011540772.1 18 - 19 2753 83
XM_017002913.1 XP_016858402.1 18 - 19 2753 83
XM_017002914.1 XP_016858403.1 18 - 19 2753 83
GenBank mRNA AB007914.2 - 16 - 17 2415 83
BC126911.1 - 15 - 16 2297 83
BC126912.1 - 15 - 16 2297 83
DQ139275.1 - 5 - 6 2133 83

View Full Product Details