Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRCIN1
Entrez Gene ID
80725
Gene Name
SRC kinase signaling inhibitor 1
Gene Aliases
P140, SNIP
Chromosome Location
Chr.17: 38530015 - 38607166 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025248.2 NP_079524.2 12 - 13 2563 88
XM_017025169.1 XP_016880658.1 13 - 14 2875 88
XM_017025170.1 XP_016880659.1 13 - 14 2730 88
XM_017025171.1 XP_016880660.1 13 - 14 2730 88
XM_017025172.1 XP_016880661.1 12 - 13 2628 88
XM_017025173.1 XP_016880662.1 13 - 14 2730 88
XM_017025174.1 XP_016880663.1 11 - 12 2170 88
GenBank mRNA AB051471.1 - 6 - 7 1075 88
AB127405.1 - 12 - 13 2563 88
AK126665.1 - 8 - 9 2273 88
AK126676.1 - 8 - 9 1700 88
AK295027.1 - 11 - 12 2170 88
BC033233.1 - 3 - 4 720 88

View Full Product Details