Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RMND5A
Entrez Gene ID
64795
Gene Name
required for meiotic nuclear division 5 homolog A
Gene Aliases
CTLH, GID2, GID2A, RMD5, p44CTLH
Chromosome Location
Chr.2: 86720291 - 86778041 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022780.3 NP_073617.1 2 - 3 658 64
GenBank mRNA AK022815.1 - 2 - 3 635 64
BC012165.1 - 2 - 3 379 64
BC047668.1 - 2 - 3 588 64

View Full Product Details